missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmin Mouse anti-Human, Bovine, Canine, Equine, Feline, Clone: DES/3255, Novus Biologicals™
Mouse Monoclonal Antibody
250.00€ - 510.00€
Specifications
| Antigen | Desmin |
|---|---|
| Clone | DES/3255 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18384572
|
Novus Biologicals
NBP3-13866-20UG |
20 μg |
250.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18321986
|
Novus Biologicals
NBP3-13866-100UG |
100 μg |
510.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Desmin Monoclonal antibody specifically detects Desmin in Human, Bovine, Canine, Equine, Feline samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Desmin | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers | |
| 10 mM PBS with 0.05% BSA | |
| CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
| Recombinant full-length human desmin protein (Uniprot: P17661 ) | |
| Primary | |
| Store at 4°C. |
| DES/3255 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Bovine, Canine, Equine, Feline | |
| P17661 | |
| 1674 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title