missing translation for 'onlineSavingsMsg'
Learn More

Desoxyanisoin, 98%

Product Code. 11419020
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11419020

Brand: Thermo Scientific Alfa Aesar A12816.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 120-44-5 | C16H16O3 | 256.301 g/mol

Deoxyanisoin react to produce a-bromo-4,4'-dimethoxy-deoxybenzoin, and this reaction could happen in the reagent of CCl4 and bromine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Deoxyanisoin react to produce a-bromo-4,4′-dimethoxy-deoxybenzoin, and this reaction could happen in the reagent of CCl4 and bromine.

Solubility
Sparingly soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-44-5
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD00008406
InChI Key SICBLYCPRWNHHP-UHFFFAOYSA-N
Synonym desoxyanisoin, deoxyanisoin, 1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxydeoxybenzoin, deoxy-p-anisoin, deoxy-4-anisoin, 1,2-bis 4-methoxyphenyl ethan-1-one, ethanone, 1,2-bis 4-methoxyphenyl, 4-methoxybenzyl 4-methoxyphenyl ketone, acetophenone, 4'-methoxy-2-p-methoxyphenyl
PubChem CID 67120
IUPAC Name 1,2-bis(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)OC

Specifications

Melting Point 109°C to 113°C
Quantity 25 g
Beilstein 1536594
Solubility Information Sparingly soluble in water.
Formula Weight 256.3
Percent Purity 98%
Chemical Name or Material Deoxyanisoin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.