missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone-d5 Acid Ethyl Ester, TRC

Product Code. 30360336
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360336 50 mg 50mg
30354493 25 mg 25mg
30358898 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30360336 Supplier Toronto Research Chemicals Supplier No. TRCD29877250MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dexamethasone acid ethyl ester ethyl-d5
Molecular Formula C23 D5 H26 F O5
Quantity 50 mg
InChI Formula InChI=1S/C23H31FO5/c1-5-29-19(27)23(28)13(2)10-17-16-7-6-14-11-15(25)8-9-20(14,3)22(16,24)18(26)12-21(17,23)4/h8-9,11,13,16-18,26,28H,5-7,10,12H2,1-4H3/t13-,16+,17+,18+,20+,21+,22+,23+/m1/s1/i1D3,5D2
SMILES [2H]C([2H])([2H])C([2H])([2H])OC(=O)[C@@]1(O)[C@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
IUPAC Name 1,1,2,2,2-pentadeuterioethyl (8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carboxylate
Molecular Weight (g/mol) 411.52
Formula Weight 411.2469
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.