missing translation for 'onlineSavingsMsg'
Learn More

Di-n-nonyl sulfide, 98%, Thermo Scientific™

Product Code. 11382916
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382916 5 g 5g
11392916 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382916 Supplier Thermo Scientific Alfa Aesar Supplier No. L10638.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 929-98-6
Molecular Formula C18H38S
Molecular Weight (g/mol) 286.56
MDL Number MFCD00027342
InChI Key KMKSVAGOBVUFRO-UHFFFAOYSA-N
Synonym dinonyl sulfide, di-n-nonyl sulfide, nonyl sulfide, 1-nonylsulfanyl nonane, nonylthiononane, dinonylsulfide, n-nonylsulfide, dinonyl sulphide, n-nonyl sulfide, octylmethylsulfide
PubChem CID 525006
IUPAC Name 1-nonylsulfanylnonane
SMILES CCCCCCCCCSCCCCCCCCC

Specifications

Melting Point 15°C
Density 0.855
Boiling Point 165°C to 167°C (3 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Linear Formula CH3(CH2)8S(CH2)8CH3
Refractive Index 1.464
Quantity 5 g
Beilstein 1755874
Formula Weight 286.56
Percent Purity 98%
Chemical Name or Material Di-n-nonyl sulfide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.