missing translation for 'onlineSavingsMsg'
Learn More

Diaveridine Hydrochloride, TRC

Product Code. 30356687
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30356687 10 mg 10mg
30356028 1 g 1g
30365624 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30356687 Supplier Toronto Research Chemicals Supplier No. TRCD12013510MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2507-23-5
Molecular Formula C13H16N4O2 . HCl
Molecular Weight (g/mol) 296.75
Synonym 5-[(3,4-Dimethoxybenzyl)pyrimidine-2,4-diamine Hydrochloride
IUPAC Name 5-[(3,4-dimethoxyphenyl)methyl]pyrimidine-2,4-diamine;hydrochloride
SMILES Cl.COc1ccc(Cc2cnc(N)nc2N)cc1OC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H16N4O2.ClH/c1-18-10-4-3-8(6-11(10)19-2)5-9-7-16-13(15)17-12(9)14;/h3-4,6-7H,5H2,1-2H3,(H4,14,15,16,17);1H
Formula Weight 296.104
Chemical Name or Material Diaveridine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.