missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl sulfide, 99%, Thermo Scientific™

Product Code. 11486046
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486046 25 g 25g
11496046 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486046 Supplier Thermo Scientific Alfa Aesar Supplier No. B22337.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-74-9
Molecular Formula C14H14S
Molecular Weight (g/mol) 214.33
MDL Number MFCD00004784
InChI Key LUFPJJNWMYZRQE-UHFFFAOYSA-N
Synonym dibenzyl sulfide, benzyl sulfide, dibenzyl sulphide, benzyl thioether, benzyl monosulfide, dibenzyl monosulfide, dibenzyl thioether, sulfide, dibenzyl, 1,3-diphenyl-2-thiapropane, benzene, 1,1'-thiobis methylene bis
PubChem CID 10867
IUPAC Name benzylsulfanylmethylbenzene
SMILES C(SCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 47°C to 50°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1911157
Merck Index 14,1145
Formula Weight 214.33
Percent Purity 99%
Chemical Name or Material Dibenzyl sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.