missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Ether, TRC

Product Code. 30379202
Change view
Click to view available options
Quantity:
50 mL
250 mL
500 mL
Unit Size:
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379202 500 mL 500mL
30378816 250 mL -
30382989 50 mL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30379202 Supplier Toronto Research Chemicals Supplier No. TRCD429465500ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-96-1
Molecular Formula C8 H18 O
Molecular Weight (g/mol) 130.23
Synonym Butyl ether (8CI), 1,1'-Oxybis[butane], Butyl oxide, Di-n-butyl ether, Dibutyl ether, Dibutyl oxide, NSC 8459, n-Bu2O, n-Butyl ether
IUPAC Name 1-butoxybutane
SMILES CCCCOCCCC

Specifications

Quantity 500 mL
InChI Formula InChI=1S/C8H18O/c1-3-5-7-9-8-6-4-2/h3-8H2,1-2H3
Formula Weight 130.1358
Chemical Name or Material Di-n-butylether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.