missing translation for 'onlineSavingsMsg'
Learn More

Dichlorotriphenylphosphorane, TRC

Product Code. 30379024
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379024 10 g 10g
30378520 5 g -
30381819 50 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30379024 Supplier Toronto Research Chemicals Supplier No. TRCD43684010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2526-64-9
Molecular Formula C18H15Cl2P
Molecular Weight (g/mol) 333.19
Synonym Triphenylphosphine Dichloride
SMILES ClP(Cl)(c1ccccc1)(c2ccccc2)c3ccccc3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C18H15Cl2P/c19-21(20,16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H
Formula Weight 332.0288
Chemical Name or Material Dichlorotriphenylphosphorane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.