missing translation for 'onlineSavingsMsg'
Learn More

Dideiodo Amiodarone, TRC

Product Code. 30012149
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012149 50 mg 50mg
30011420 100 mg 100mg
30007794 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012149 Supplier Toronto Research Chemicals Supplier No. TRCD43942050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23551-25-9
Molecular Formula C25 H31 N O3
Molecular Weight (g/mol) 393.52
Synonym Ketone, 2-butyl-3-benzofuranyl p-[2-(diethylamino)ethoxy]phenyl (7CI,8CI), Deiodoamiodarone, L 3937, (2-Butylbenzo-furan-3-yl)[4-[2-(diethylamino)ethoxy]phenyl]-methanone, Amiodaronhydrochorid Impurity A
IUPAC Name (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]phenyl]methanone
SMILES CCCCc1oc2ccccc2c1C(=O)c3ccc(OCCN(CC)CC)cc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C25H31NO3/c1-4-7-11-23-24(21-10-8-9-12-22(21)29-23)25(27)19-13-15-20(16-14-19)28-18-17-26(5-2)6-3/h8-10,12-16H,4-7,11,17-18H2,1-3H3
Formula Weight 393.2304
Percent Purity >95%
Chemical Name or Material Dideiodo Amiodarone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.