missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (2-pentyl)malonate, 94%, Thermo Scientific™

Product Code. 11471577
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471577 250 g 250g
11324509 10 g 10g
11451577 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471577 Supplier Thermo Scientific Alfa Aesar Supplier No. B24993.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Diethyl (2-pentyl)malonate is used in vitro reaction of barbiturates with formaldehyde to evaluate forensic toxicology on the cause of death.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-47-5
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.30
MDL Number MFCD00051558
InChI Key RQFSNEWORATSCC-UHFFFAOYNA-N
Synonym diethyl 1-methylbutyl malonate, diethyl 2-pentyl malonate, diethyl 1-methylbutylmalonate, diethyl 1-methylbutal malonate, 1-methylbutyl malonic ester, 1-methylbutyl malonic acid diethyl ester, diethyl pentan-2-ylmalonate, acmc-1brsl, diethyl sec-amyl malonate
PubChem CID 95449
IUPAC Name diethyl 2-pentan-2-ylpropanedioate
SMILES CCCC(C)C(C(=O)OCC)C(=O)OCC

Specifications

Density 0.968
Boiling Point 118°C to 119°C (9 mmHg)
Refractive Index 1.427
Quantity 250 g
Formula Weight 230.31
Percent Purity 94%
Chemical Name or Material Diethyl (2-pentyl)malonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.