missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Phthalate-13C6, TRC

Product Code. 30387873
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387873 5 mg 5mg
30375880 50 mg 50mg
30376254 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387873 Supplier Toronto Research Chemicals Supplier No. TRCD4447735MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Phthalate-13C6
Molecular Formula 13C6 C6 H14 O4
Quantity 5 mg
Synonym 1,2-Benzenedicarboxylic Acid 1,2-Diethyl Ester-13C6, 1,2-Benzenedicarboxylic Acid Diethyl Ester-13C6, Phthalic Acid Diethyl Ester-13C6, Anozol-13C6, DEP-13C6, Diethyl 1,2-Benzenedicarboxylate-13C6, Diethyl Phthalate-13C6, Ethyl Phthalate-13C6, NSC 8905-13C6, Neantine-13C6, Palatinol A-13C6, Phthalol-13C6, Placidol E-13C6, Solvanol-13C6, Unimoll DA-13C6, o-Benzenedicarboxylic Acid Diethyl Ester-13C6, o-Bis(ethoxycarbonyl)benzene-13C6
InChI Formula InChI=1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3/i5+1,6+1,7+1,8+1,9+1,10+1
SMILES CCOC(=O)[13c]1[13cH][13cH][13cH][13cH][13c]1C(=O)OCC
IUPAC Name diethyl (1,2,3,4,5,6-13C6)cyclohexa-2,4,6-triene-1,2-dicarboxylate
Molecular Weight (g/mol) 228.193
Formula Weight 228.109
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.