missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Pyrocarbonate, Serva Electrophoresis™

Product Code. p-8014567
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11701987 10 mL Each
11417573 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11701987 Supplier Serva Electrophoresis Supplier No. 1883501
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1609-47-8
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00009106
InChI Key FFYPMLJYZAEMQB-UHFFFAOYSA-N
Synonym diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
PubChem CID 3051
ChEBI CHEBI:59051
IUPAC Name diethyl dicarbonate
SMILES CCOC(=O)OC(=O)OCC

Specifications

Refractive Index 1.3960 - 1.4000
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.