missing translation for 'onlineSavingsMsg'
Learn More

Diiron nonacarbonyl, 98%

Product Code. 10388150 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10388150 10 g 10g
10398150 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10388150 Supplier Thermo Scientific Acros Supplier No. 209280100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 15321-51-4 | C9H3Fe2O9 | 366.804 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15321-51-4
Molecular Formula C9H3Fe2O9
MDL Number MFCD00151465
InChI Key CGJSNKHFIBINMR-UHFFFAOYSA-N
Synonym iron nonacarbonyl, enneacarbonyldiiron, nonacarbonyldiiron, 1,3-diferrabicyclo 1.1.1 pentane-2,4,5-trione; hexakis carbon monoxide, diironnonacarbonyl, diironnonacarbonyl, purum fe
PubChem CID 71311424

Specifications

Color Black to Brown-Red
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle packed in tin can
Linear Formula Fe2(CO)9
Quantity 10 g
Fieser 01,259; 02,139; 03,101; 04,157; 05,221; 06,195; 07,110; 08,498; 13,320; 04,534; 05,706; 07,144; 08,41; 08,216; 10,174; 08,214
SMILES [CH-]=O.[CH-]=O.[CH-]=O.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Fe].[Fe+3]
IUPAC Name carbon monoxide;iron;iron(3+);methanone
Molecular Weight (g/mol) 366.804
Formula Weight 363.78
Percent Purity ≥97.5%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Diiron nonacarbonyl, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Diiron nonacarbonyl
Signal Word
  • Danger
Hazard Category
  • Pyrophoric solid Category 1
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P243-Take precautionary measures against static discharge.
  • P422-Store contents under .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.