missing translation for 'onlineSavingsMsg'
Learn More

Diisononyl Phthalate and Diisodecyl Phthalate Mixture 391 1000 μg/mL in Isooctane, Dr. Ehrenstorfer

Product Code. 30321453
Change view
Click to view available options
Quantity:
5x1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321453 5x1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321453 Supplier Dr. Ehrenstorfer Supplier No. DREGS09000391IO
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diisononyl Phthalate (mixture of branched chain isomers)∣Phthalic acid, bis-isodecyl ester
CAS 68515-48-0, 26761-40-0
Quantity 5x1 mL
Concentration or Composition (by Analyte or Components) Diisononyl Phthalate (mixture of branched chain isomers) 1000 μg/mL∣Phthalic acid, bis-isodecyl ester 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.