missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxymethane, Pure, Fisher Chemical

Product Code. 10325800 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber glass bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10325800 500 mL Amber glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10325800 Supplier Fisher Chemical Supplier No. D/3400/08

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 109-87-5
Melting Point -105°C
Color Colorless
Boiling Point 41°C
Packaging Amber glass bottle
Molecular Formula C3H8O2
MDL Number 8495
Quantity 500 mL
Synonym methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether
InChI Key NKDDWNXOKDWJAK-UHFFFAOYSA-N
SMILES COCOC
IUPAC Name dimethoxymethane
Molecular Weight (g/mol) 76.095
PubChem CID 8020
ChEBI CHEBI:48341
Formula Weight 76.09g/mol
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Dimethoxymethane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.