missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (2-thienylmethyl)malonate, 95%, Thermo Scientific™

Product Code. 15493437
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493437 250 mg 250mg
11401160 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493437 Supplier Thermo Scientific Alfa Aesar Supplier No. H26301.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122308-25-2
Molecular Formula C10H12O4S
Molecular Weight (g/mol) 228.26
MDL Number MFCD08436111
InChI Key PIIHAJHYNTWOAF-UHFFFAOYSA-N
Synonym dimethyl 2-thienylmethyl malonate, dimethyl 2-thiophen-2-ylmethyl malonate, 1,3-dimethyl 2-thiophen-2-ylmethyl propanedioate, acmc-1c16u, 2-thienylmethyl malonic acid dimethyl ester, dimethyl thiophen-2-yl methyl propanedioate, propanedioic acid,2-2-thienylmethyl-, 1,3-dimethyl ester
PubChem CID 14425092
IUPAC Name dimethyl 2-(thiophen-2-ylmethyl)propanedioate
SMILES COC(=O)C(CC1=CC=CS1)C(=O)OC

Specifications

Quantity 250 mg
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 228.27
Percent Purity 95%
Chemical Name or Material Dimethyl (2-thienylmethyl)malonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.