missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl octafluoroadipate, 94%, Thermo Scientific™

Product Code. 11379375
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379375 5 g 5g
11389375 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379375 Supplier Thermo Scientific Alfa Aesar Supplier No. L16752.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3107-98-0
Molecular Formula C8H6F8O4
Molecular Weight (g/mol) 318.119
MDL Number MFCD00042267
InChI Key XPXVIIILXUOEQA-UHFFFAOYSA-N
Synonym dimethyl octafluoroadipate, dimethyl perfluoroadipate, hexanedioic acid, octafluoro-, dimethyl ester, 1,6-dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate, dimethyloctafluoroadipate, acmc-1cson, dimethyl octafluoroadipinate, dimethyl octafluorohexanedioate, perfluoroadipic acid dimethyl ester, octafluoroadipic acid dimethyl ester
PubChem CID 2737117
IUPAC Name dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate
SMILES COC(=O)C(C(C(C(C(=O)OC)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.547
Boiling Point 120°C (15 mmHg)
Linear Formula CH3O2C(CF2)4CO2CH3
Refractive Index 1.3475
Quantity 5 g
Beilstein 1916112
Formula Weight 318.12
Percent Purity 94%
Chemical Name or Material Dimethyl octafluoroadipate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.