missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl tert-butylmalonate, 95%

Product Code. 11475940
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475940 1 g 1g
11485940 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475940 Supplier Thermo Scientific Alfa Aesar Supplier No. H26532.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 39520-25-7 | C9H16O4 | 188.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39520-25-7
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD08436110
InChI Key OBANUQSDPDUGSL-UHFFFAOYSA-N
PubChem CID 13574047
IUPAC Name dimethyl 2-tert-butylpropanedioate
SMILES COC(=O)C(C(=O)OC)C(C)(C)C

Specifications

Refractive Index 1.428
Quantity 1 g
Synonym dimethyl 2-tert-butyl malonate, dimethyl tert-butylmalonate, propanedioic acid, 1,1-dimethylethyl-, dimethyl ester, 1,3-dimethyl 2-tert-butylpropanedioate, acmc-1aijh, dimethyl2-tert-butyl malonate, dimethyl tert-butylpropanedioate, tert-butylmalonic acid dimethyl ester, propanedioic acid,2-1,1-dimethylethyl-, 1,3-dimethyl ester
Formula Weight 188.22
Percent Purity 95%
Chemical Name or Material Dimethyl tert-butylmalonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.