missing translation for 'onlineSavingsMsg'
Learn More

Diphenhydramine Hydrochloride (1.0 mg/mL in Methanol), TRC

Product Code. 30403827
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403827 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403827 Supplier Toronto Research Chemicals Supplier No. TRCKIT120510X1ML
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 147-24-0
Molecular Formula C17 H21 N O . Cl H
Quantity 10 x 1 mL
Synonym Diphenhydramine Hydrochloride, Orphenadrine Hydrochloride Imp. D (EP) as Hydrochloride, Ethanamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (1:1), Ethanamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (9CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8CI), β-Dimethylaminoethyl benzhydryl ether hydrochloride, 2-(Benzhydryloxy)-N,N-dimethylethylamine hydrochloride, 2-Diphenylmethoxy-N,N-dimethylethylamine hydrochloride, Alledryl, Allergan, Allergina, Allergival, Amidryl, Asdrin, Bagodryl, Bax, Bena, Bena-Fedrin, Benadril hydrochloride, Benadryl, Benocten, Benodine, Benylin, Benzantin, Benzhydramine hydrochloride, Benzhydrol β-dimethylaminoethyl ether hydrochloride, Denydryl, Dibondrin, Difenhydramine hydrochloride, Dimedrol, Dimedrol hydrochloride, Diphantine, Fenylhist, Halbmond, Histacyl, Noctomin, Resmin, Restamin, S 8, Sedopretten, Sekundal D, Syntedril, Vena, Wehydryl
InChI Formula InChI=1S/C17H21NO.ClH/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;/h3-12,17H,13-14H2,1-2H3;1H
SMILES Cl.CN(C)CCOC(c1ccccc1)c2ccccc2
IUPAC Name 2-benzhydryloxy-N,N-dimethylethanamine;hydrochloride
Molecular Weight (g/mol) 291.82
Formula Weight 291.139
Concentration or Composition (by Analyte or Components) Diphenhydramine hydrochloride 1 mg/mL
Chemical Name or Material Diphenhydramine hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.