missing translation for 'onlineSavingsMsg'
Learn More

Diphenhydramine Hydrochloride, TRC

Product Code. 30135368
Change view
Click to view available options
Quantity:
100 mg
250 mg
5 g
Unit Size:
100mg
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135368 250 mg 250mg
17953855 5 g 5g
17913175 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135368 Supplier Toronto Research Chemicals Supplier No. TRCD486900250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-24-0
Molecular Formula C17 H21 N O . Cl H
Molecular Weight (g/mol) 291.82
Synonym Diphenhydramine Hydrochloride, Orphenadrine Hydrochloride Imp. D (EP) as Hydrochloride, Ethanamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (1:1), Ethanamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (9CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8CI), β-Dimethylaminoethyl benzhydryl ether hydrochloride, 2-(Benzhydryloxy)-N,N-dimethylethylamine hydrochloride, 2-Diphenylmethoxy-N,N-dimethylethylamine hydrochloride, Alledryl, Allergan, Allergina, Allergival, Amidryl, Asdrin, Bagodryl, Bax, Bena, Bena-Fedrin, Benadril hydrochloride, Benadryl, Benocten, Benodine, Benylin, Benzantin, Benzhydramine hydrochloride, Benzhydrol β-dimethylaminoethyl ether hydrochloride, Denydryl, Dibondrin, Difenhydramine hydrochloride, Dimedrol, Dimedrol hydrochloride, Diphantine, Fenylhist, Halbmond, Histacyl, Noctomin, Resmin, Restamin, S 8, Sedopretten, Sekundal D, Syntedril, Vena, Wehydryl
IUPAC Name 2-benzhydryloxy-N,N-dimethylethanamine;hydrochloride
SMILES Cl.CN(C)CCOC(c1ccccc1)c2ccccc2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H21NO.ClH/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;/h3-12,17H,13-14H2,1-2H3;1H
Formula Weight 291.139
Percent Purity >95%
Chemical Name or Material Diphenhydramine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.