missing translation for 'onlineSavingsMsg'
Learn More

Diphenolic Acid, TRC

Product Code. 30358377
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358377 10 g 10g
30364960 5 g 5g
30368655 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358377 Supplier Toronto Research Chemicals Supplier No. TRCD26335010G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-00-1
Molecular Formula C17H18O4
Molecular Weight (g/mol) 286.32
Synonym 4,4-Bis(4-hydroxyphenyl)pentanic Acid, 4,4-Bis(4-hydroxyphenyl)pentanoic Acid, 4,4-Bis(4-hydroxyphenyl)valeric Acid, 4,4-Bis(p-hydroxyphenyl)pentanoic Acid, 4,4-Bis(p-hydroxyphenyl)valeric Acid, 4,4'-Bis(4-hydroxyphenyl)valeric Acid, Bisphenol Acid, D 1274, DPA, Diphenolic Acid, NSC 3371, NSC 34824, NSC 55069, gamma,gamma-Bis(p-hydroxyphenyl)valeric Acid
SMILES CC(CCC(=O)O)(c1ccc(O)cc1)c2ccc(O)cc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C17H18O4/c1-17(11-10-16(20)21,12-2-6-14(18)7-3-12)13-4-8-15(19)9-5-13/h2-9,18-19H,10-11H2,1H3,(H,20,21)
Formula Weight 286.1205
Chemical Name or Material Diphenolic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.