Learn More
Diphenylacetyl chloride, 97.5%
Product Code. 10264951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10264951 25 g Glass bottle 25g
10375221 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10264951 Supplier Thermo Scientific Acros Supplier No. 292020250
Request Bulk or Custom Format
CAS: 1871-76-7 | C14H11ClO | 230.691 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1871-76-7
MDL Number MFCD00013655
InChI Key MSYLETHDEIJMAF-UHFFFAOYSA-N
Synonym diphenylacetyl chloride, diphenylacetic acid chloride, diphenylacetylchloride, acetyl chloride, diphenyl, unii-6553c1cb9s, alpha-phenylbenzeneacetyl chloride, diphenyl-acetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, a-phenylbenzeneacetyl chloride, diphenyl acetyl chloride
PubChem CID 74637
IUPAC Name 2,2-diphenylacetyl chloride
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl
Specifications
Melting Point 51°C to 56°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5%
Packaging Glass bottle
Molecular Formula C14H11ClO
Linear Formula (C6H5)2CHCOCl
Quantity 25 g
Beilstein 09, 674
Merck Index 15, 3346
Molecular Weight (g/mol) 230.691
Formula Weight 230.69
Percent Purity ≥96.5%
Physical Form Crystalline Powder
Chemical Name or Material Diphenylacetyl chloride, 97.5%