missing translation for 'onlineSavingsMsg'
Learn More

Diuron-d6 (dimethyl-d6), CDN

Product Code. 30322238
Change view
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322238 0.05 g 0.05g
30313850 0.01 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30322238 Supplier CDN Supplier No. CDND56440.05G
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diuron D6
CAS 1007536-67-5
Molecular Formula C9 2H6 H4 Cl2 N2 O
Quantity 0.05 g
Synonym Urea, N'-(3,4-dichlorophenyl)-N,N-di(methyl-d3)-, N'-(3,4-Dichlorophenyl)-N,N-di(methyl-d3)urea, Diuron-D6, Diuron-D6 (dimethyl-D6)
InChI Formula InChI=1 S/C9H10Cl2N2O/c1-13(2)9(14)12-6-3-4-7(10)8(11)5-6/h3-5 H,1-2H3,(H,12,14)/i1D3,2D3
SMILES [2 H]C([2 H])([2 H])N(C(=O)Nc1ccc(Cl)c(Cl)c1)C([2 H])([2 H])[2 H]
IUPAC Name 3-(3,4-dichlorophenyl)-1,1-bis(trideuteriomethyl)urea
Molecular Weight (g/mol) 239.13
Formula Weight 238.0547 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.