missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase iota Antibody (OTI5H9), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44868-100ul
This item is not returnable.
View return policy
Description
DNA Polymerase iota Monoclonal antibody specifically detects DNA Polymerase iota in Human samples. It is validated for Western Blot
Specifications
| DNA Polymerase iota | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5H9 | |
| Western Blot 1:2000 | |
| DNA polymerase iota, EC 2.7.7.7, Eta2, polymerase (DNA directed) iota, RAD30 homolog B, RAD30Bpolymerase (DNA-directed), iota, RAD3OB | |
| Human recombinant protein fragment corresponding to amino acids 458-740 of human POLI (NP_009126) produced in E.coli. | |
| 100 μL | |
| Chromatin Research, DNA Polymerases, DNA Repair | |
| 11201 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction