Learn More
DNA/RNA Damage Antibody (15A3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 12 publications
Brand: Novus Biologicals NB110-96878
This item is not returnable.
View return policy
Description
DNA/RNA Damage Monoclonal specifically detects DNA/RNA Damage in Human, Mouse, Non-species specific samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoassay, Dot Blot, Functional Assay.
Specifications
| DNA/RNA Damage | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Immunoassay, Dot Blot 1:100-1:200, Functional Assay | |
| 8-hydroxy-guanosine-BSA and-casein conjugates | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Non-species specific | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, Dot Blot, Functional Assay | |
| 15A3 | |
| Unconjugated | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Recognizes markers of oxidative damage to DNA (8-hydroxy-2'-deoxyguanosine, 8-hydroxyguanine and 8-hydroxyguanosine). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |