Learn More
DNA/RNA Damage Antibody (15A3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 12 publications
Brand:  Novus Biologicals NB110-96878
 View more versions of this product
Product Code. 18731694
  • 644.00€ / 0.10mg
Description
DNA/RNA Damage Monoclonal specifically detects DNA/RNA Damage in Human, Mouse, Non-species specific samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoassay, Dot Blot, Functional Assay.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNA/RNA Damage
Monoclonal
1 mg/mL
Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Immunoassay, Dot Blot 1:100-1:200, Functional Assay
8-hydroxy-guanosine-BSA and-casein conjugates
0.1 mg
Primary
Human, Mouse, Non-species specific
Purified
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, Dot Blot, Functional Assay
15A3
Unconjugated
Mouse
Protein G purified
RUO
Recognizes markers of oxidative damage to DNA (8-hydroxy-2'-deoxyguanosine, 8-hydroxyguanine and 8-hydroxyguanosine).
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2b