missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA/RNA Damage Antibody (15A3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 12 publications
258.00€ - 644.00€
Specifications
| Antigen | DNA/RNA Damage |
|---|---|
| Clone | 15A3 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Immunoassay, Dot Blot 1:100-1:200, Functional Assay |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, Dot Blot, Functional Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18045224
|
Novus Biologicals
NB110-96878SS |
0.025 mg |
258.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18731694
|
Novus Biologicals
NB110-96878 |
0.1 mg |
644.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNA/RNA Damage Monoclonal specifically detects DNA/RNA Damage in Human, Mouse, Non-species specific samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoassay, Dot Blot, Functional Assay.Specifications
| DNA/RNA Damage | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, Dot Blot, Functional Assay | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Non-species specific | |
| IgG2b | |
| Protein G purified | |
| Recognizes markers of oxidative damage to DNA (8-hydroxy-2'-deoxyguanosine, 8-hydroxyguanine and 8-hydroxyguanosine). |
| 15A3 | |
| Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Immunoassay, Dot Blot 1:100-1:200, Functional Assay | |
| Monoclonal | |
| Purified | |
| RUO | |
| 8-hydroxy-guanosine-BSA and-casein conjugates | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title