missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecanoic-2,2-d2 Acid, CDN
Specifications
Specifications
| Chemical Name or Material | Dodecanoic-2,2-D2 Acid |
| CAS | 64118-39-4 |
| Molecular Formula | C12 D2 H22 O2 |
| Quantity | 0.5 g |
| Synonym | Dodecanoic-2,2-d2 acid (9 CI), Lauric-2,2-d2 acid (6 CI, 7 CI), 2,2-Dideuteriododecanoic acid, α-Dideuterododecanoic acid, Dodecanoic acid-2,2-d2, Lauric acid-2,2-d2, [2,2-2H2]Dodecanoic acid, Dodecanoic-2,2-D2 acid |
| InChI Formula | InChI=1 S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)/i11D2 |
| SMILES | [2 H]C([2 H])(CCCCCCCCCC)C(=O)O |
| IUPAC Name | 2,2-dideuteriododecanoic acid |
| Molecular Weight (g/mol) | 202.3301 |
| Formula Weight | 202.1902 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?