missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DOK2 Monoclonal Antibody (OTI2D10)
GREENER_CHOICE

Product Code. 15915264
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15915264 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15915264 Supplier Invitrogen™ Supplier No. MA525661

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DOK2 Monoclonal Antibody for Western Blot

The protein encoded by this gene is constitutively tyrosine phosphorylated in hematopoietic progenitors isolated from chronic myelogenous leukemia patients in the chronic phase. It may be a critical substrate for p210(bcr/abl), a chimeric protein whose presence is associated with CML. This encoded protein binds p120 from CML cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOK2
Applications Western Blot
Classification Monoclonal
Clone OTI2D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DOK2
Gene Accession No. O60496, O70469
Gene Alias docking protein 2; docking protein 2, 56kD; docking protein 2, 56kDa; Dok2; Dok-2; DokR; Dok-R; dok-related protein; Downstream of tyrosine kinase 2; Frip; IL-Four Receptor Interacting Protein; IL-four receptor-interacting protein; LOW QUALITY PROTEIN: docking protein 2; docking protein 2; p56 dok-2; p56(dok-2); p56DOK; p56dok-2
Gene Symbols DOK2
Host Species Mouse
Immunogen Full length human recombinant protein of DOK2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13449, 290361, 486133, 9046
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.