Learn More
DR6 Mouse anti-Human, Clone: 3C2D10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66754-1-IG-20UL
This item is not returnable.
View return policy
Description
DR6 is widely expressed in human tissues and cell lines. The ligand for DR6 has not been identified. Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNF-R1 and Fas. Several novel death receptors including DR3, DR4, and DR5 were recently identified. A new death domain containing receptor in the TNFR family was cloned recently and termed DR6 for death receptor-6. Like TNF-R1, DR6 interacts with death domain containing adapter molecule TRADD. Overexpression of DR6 induces apoptosis and activates NF-kappa-B and JNK.Specifications
| DR6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75509 | |
| TNFRSF21 | |
| DR6 Fusion Protein Ag14327 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 3C2D10 | |
| Unconjugated | |
| TNFRSF21 | |
| BM 018, CD358, Death receptor 6, DR6, TNFRSF21 | |
| Mouse | |
| Protein G | |
| RUO | |
| 27242 | |
| -20°C | |
| Liquid |