missing translation for 'onlineSavingsMsg'
Få mere at vide

DSPP Antibody - Azide and BSA Free, Novus Biologicals™

Artikelnummer. 18645842 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
0.02 mL
0.1 mL
Pakningsstørrelse:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
18645842 0.02 mL 0.02mL
18608052 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18645842 Leverandør Novus Biologicals Leverandørnr. NBP2925460.02ml

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

DSPP Polyclonal antibody specifically detects DSPP in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen DSPP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias dentin phosphoprotein, dentin sialophosphoprotein, dentin sialoprotein, DFNA39, DMP3, DTDP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 16-146 of human DSPP (NP_055023.2). IPVPQSKPLERHVEKSMNLHLLARSNVSVQDELNASGTIKESGVLVHEGDRGRQENTQDGHKGEGNGSKWAEVGGKSFSTYSTLANEEGNIEGWNGDTGKAETYGHDGIHGKEENITANGIQGQVSIIDNA
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1834
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.