missing translation for 'onlineSavingsMsg'
Learn More

DSS1 Antibody [DyLight 594], Novus Biologicals™

Product Code. 30613464 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30613464 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30613464 Supplier Novus Biologicals Supplier No. NBP346629DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DSS1 Polyclonal antibody specifically detects DSS1 in Human samples. It is validated for ELISA,Immunohistochemistry,Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DSS1
Applications ELISA, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias 26S proteasome complex subunit DSS1, Deleted in split hand/split foot protein 1, deleted in split-hand/split-foot 1, DSS1Split hand/foot malformation type 1 protein, ECD, SEM1, Shfdg1, SHSF1SHFD1, Split hand/foot deleted protein 1, split hand/foot malformation (ectrodactyly) type 1
Host Species Rabbit
Immunogen split hand/foot malformation(ectrodactyly) type 1. (Uniprot# P60896)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7979
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.