Learn More
DWK Life Sciences DURAN™ Bell Jar, with glass knob top
Product Code. 10304961 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Base:
185 mm
260 mm
315 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Base unitSize
10304961 260 mm Each
12739666 185 mm Each
12759666 315 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10304961 Supplier DWK Life Sciences Supplier No. 244606607
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
Flat flange bell with knob and ground flange for mounting on a vacuum experimental plate (vacuum chamber).
By default, tests are carried out at normal pressure. The conduct of tests under vacuum (negative pressure) can have significant effects on the reaction sequence. In order to be able to observe these effects in the rough and fine vacuum range, a vacuum experiment plate and a bell jar (also called air pump bell) are required.
  • The button makes it easy to raise the bell jar
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Base 260 mm
Material Borosilicate Glass (3.3)
For Use With (Application) Vacuum applications; Tests in vacuum chambers
No. per Pack 1/Pk.