Learn More
DWK Life Sciences DURAN™ Separating Funnel, conical shape, with glass key
Product Code. 10763121 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10763121 100 mL Each
10379600 250 mL Each
10470021 500 mL Each
10359650 1000 mL Each
10369650 2000 mL Each
10516532 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10763121 Supplier DWK Life Sciences Supplier No. 242942409
  • ONLINE EXCLUSIVE PRICE
    82.27€
    71.94€ / Pack of 1
    Save 10.33€
    12% Off
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The separating funnel enables the separation of immiscible liquids (liquid-liquid extraction).
A separating funnel has the shape of a pointed flask with a drain cock at its tip. The separating funnel is used for the liquid-liquid extraction of two immiscible liquids (substance separation process) and can thus also be used for cleaning processes. The liquid with the higher density collects in the lower part of the vessel and can be drained via the drain cock. The printed scale allows an exact volume determination.
  • The conical shape makes it highly suited for phase separation
  • With standard ground stopcock, retaining device and plastic stopper
  • With solid key
  • Fired-on, highly durable white print
Specifications
Capacity (Metric) 100 mL
Diameter (Metric) Stem 9 mm
Material Boro 3.3, Polypropylene
Autoclavable Autoclavable
For Use With (Application) Phase separation; Liquid-liquid extraction
Quantity 1 Ea.