Learn More
DWK Life Sciences DURAN™ Silicone Lid, stretchable, Set
The stretchable silicone lid is used to close round and square vessels and is available in three sizes and as a set as well as in three colours each.
8.23€
Specifications
Description Autoclavable (Silicone lid only without glassware)
Material Silicone
For Use With (Application) Covering; color coding and identification
Includes 1 Piece Each Size
No. per Pack 1/Pk.
View More Specs
Products 3
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
16464944
View Documents
DWK Life Sciences
291100011
Pink
ONLINE EXCLUSIVE PRICE
8.50€
8.23€ / Each
Save 0.27€
3% Off
View alternative products
Save Up to 
»
Only null left
In Stock
Add to basket
16474944
View Documents
DWK Life Sciences
291100028
Light Blue
ONLINE EXCLUSIVE PRICE
8.50€
8.23€ / Each
Save 0.27€
3% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 01-12-2025
Add to basket
16484944
View Documents
DWK Life Sciences
291100036
Green
ONLINE EXCLUSIVE PRICE
8.50€
8.23€ / Each
Save 0.27€
3% Off
View alternative products
Save Up to 
»
Only null left
In Stock
Add to basket
Description
Description
The closure or covering of vessels is an everyday topic in the laboratory. Often glass plates or aluminum foil are used. From the point of view of environmental friendliness and durability, both variants are rather classified as disadvantageous. The newly developed silicone cover is a good alternative to conventional cover variants and is available individually in three sizes or as a set. Due to its elasticity, the lid can be used for a variety of vessel openings, square or round. The silicone lid has a slightly larger tab that can be used as a writing field. The three colors offered - cyan, pink and green - also allow color identification.
The stretchy DURAN™ silicone lids are resistant to both chemicals and heat. They are recommended for use at temperatures between -40 and +180°C. The DURAN™ lids are manufactured from silicone, which is an autoclavable, and microwave-safe material. However due to the pressure changes that occur during autoclaving the lids may become loose, or tear, as a result they are not recommended for this type of application.
  • For covering and sealing laboratory vessels
  • Stretchable and thus suitable for a large number of vessel openings
  • Available in different colors for clear differentiation and identification
  • Easy to remove with the aid of a flap, both with and without gloves
  • Chemically resistant, heat resistant and reusable
Specifications
Specifications
Autoclavable (Silicone lid only without glassware)
Covering; color coding and identification
1/Pk.
Silicone
1 Piece Each Size
Set S-M-L