Learn More
Ecdysterone, Thermo Scientific™
An insect steroid hormone that induces apoptosis
212.99€ - 709.02€
Chemical Identifiers
| CAS | 5289-74-7 |
|---|---|
| Molecular Formula | C27H44O7 |
| Molecular Weight (g/mol) | 480.64 |
| MDL Number | MFCD00036740 |
| InChI Key | NKDFYOWSKOHCCO-YPVLXUMRSA-N |
| Synonym | 20-hydroxyecdysone, ecdysterone, beta-ecdysone, crustecdysone, 20-oh ecdysone, the-7, polypodine a, insect moulting hormone, commisterone, crustecdyson |
| PubChem CID | 5459840 |
| ChEBI | CHEBI:16587 |
| IUPAC Name | (1S,3aS,5aR,7R,8S,9aR,9bR,11aR)-3a,7,8-trihydroxy-9a,11a-dimethyl-1-[(2R,3R)-2,3,6-trihydroxy-6-methylheptan-2-yl]-1H,2H,3H,3aH,5H,5aH,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-5-one |
| SMILES | CC(C)(O)CC[C@@H](O)[C@](C)(O)[C@H]1CC[C@@]2(O)C3=CC(=O)[C@@H]4C[C@@H](O)[C@@H](O)C[C@]4(C)[C@H]3CC[C@]12C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15455359
|
Thermo Scientific Alfa Aesar
J63091.MA |
10mg |
217.00€ 212.99€ / 10mg Save 4.01€ 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15465359
|
Thermo Scientific Alfa Aesar
J63091.MF |
50mg |
723.00€ 709.02€ / 50mg Save 13.98€ 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsEcdysterone is an insect steroid hormone that induces apoptosis, cell proliferation, growth by controlling gene expression involved in molting and metamorphosis. And it is also used as a ecdysteroid in both plant and animal species that controls cell death.
Solubility
Soluble in alcohol
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
| 5289-74-7 | |
| 480.64 | |
| NKDFYOWSKOHCCO-YPVLXUMRSA-N | |
| 5459840 | |
| (1S,3aS,5aR,7R,8S,9aR,9bR,11aR)-3a,7,8-trihydroxy-9a,11a-dimethyl-1-[(2R,3R)-2,3,6-trihydroxy-6-methylheptan-2-yl]-1H,2H,3H,3aH,5H,5aH,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-5-one |
| C27H44O7 | |
| MFCD00036740 | |
| 20-hydroxyecdysone, ecdysterone, beta-ecdysone, crustecdysone, 20-oh ecdysone, the-7, polypodine a, insect moulting hormone, commisterone, crustecdyson | |
| CHEBI:16587 | |
| CC(C)(O)CC[C@@H](O)[C@](C)(O)[C@H]1CC[C@@]2(O)C3=CC(=O)[C@@H]4C[C@@H](O)[C@@H](O)C[C@]4(C)[C@H]3CC[C@]12C |
Specifications
| 5289-74-7 | |
| MFCD00036740 | |
| 14,3491 | |
| Soluble in alcohol | |
| CC(C)(O)CC[C@@H](O)[C@](C)(O)[C@H]1CC[C@@]2(O)C3=CC(=O)[C@@H]4C[C@@H](O)[C@@H](O)C[C@]4(C)[C@H]3CC[C@]12C | |
| 480.64 | |
| CHEBI:16587 | |
| Ecdysterone |
| C27H44O7 | |
| 1917578 | |
| 20-hydroxyecdysone, ecdysterone, beta-ecdysone, crustecdysone, 20-oh ecdysone, the-7, polypodine a, insect moulting hormone, commisterone, crustecdyson | |
| NKDFYOWSKOHCCO-YPVLXUMRSA-N | |
| (1S,3aS,5aR,7R,8S,9aR,9bR,11aR)-3a,7,8-trihydroxy-9a,11a-dimethyl-1-[(2R,3R)-2,3,6-trihydroxy-6-methylheptan-2-yl]-1H,2H,3H,3aH,5H,5aH,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-5-one | |
| 5459840 | |
| 480.64 |
Sécurité et traitement
missing translation for 'rtecsNumber' : FZ8060000
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.