missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EDG2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11595882
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11595882 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11595882 Supplier Invitrogen™ Supplier No. PA11041

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-1041 detects lysophosphatidic acid receptor 1 protein in rat, human, mouse, and Xenopus samples. PA1-1041 has successfully been used in Western blot procedures. By Western blot, this antibody detects an ~42 kDa protein representing lysophosphatidic acid receptor 1 in PC12 and HeLa whole cell lysates as well as mouse and Xenopus liver samples. The PA1-1041 immunogen is a synthetic peptide corresponding to residues L(348) N H T I L A G V H S N D H S V V(364) of human LPA1. The immunizing sequence is completely conserved in human, mouse, rat and bovine. This peptide (Cat. # PEP-233) is available for use in neutralization and control experiments.

EDG2 belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids There are eight known members of the EDG receptor family and they are implicated in mediating growth-related effects such as induction of cellular proliferation, alterations in differentiation and survival, and suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDG2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA and 0.05% sodium azide
Gene LPAR1
Gene Accession No. P61793, P61794, Q92633
Gene Alias AI326300; Edg2; edg-2; endothelial differentiation lysophosphatidic acid G-protein-coupled receptor 2; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 2; Gpcr26; Gpcr91; GPR26; G-protein coupled receptor 26; Kdt2; LPA receptor 1; LPA1; LPA-1; LPAR1; Lysophosphatidic acid receptor 1; lysophosphatidic acid receptor Edg-2; Mrec1.3; rec.1.3; rec1.3; ventricular zone gene 1; Vzg1; vzg-1
Gene Symbols LPAR1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues L(348) N H T I L A G V H S N D H S V V(364) of human LPA1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116744, 14745, 1902
Target Species Human, Mouse, Rat, Frog
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.