missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: QALSSASKFE AELKAEQDER KREEE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
EFHD1, also known as Swiprosin-2 or SWS2, is an EF-hand and coiled-coil-containing adaptor protein identified in a subtractive hybridization study using a neuronal cell line established from the cerebellum of an adult p53-null mouse, however further study indicated no difference in normal mice. Its mRNA is widely expressed, with its expression in brain undetectable at embryonic stages, with increasing levels from postnatal to adult development. In situ hybridization showed expression in neurons but not white matter of the cerebellum and cerebrum. EHFD1 is also highly expressed in testes, ovary, and the collecting ducts of the kidney, suggesting that in non-neuronal cells, EFHD1 may be involved in gametogenesis and water-reabsorbtion.
Specifications
Specifications
| Antigen | EFHD1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | EFHD1 |
| Gene Accession No. | Q9BUP0 |
| Gene Alias | 4931430I01Rik; AI452351; EF hand domain containing 1; EF-hand domain family member D1; EF-hand domain family, member D1; EF-hand domain-containing protein 1; EF-hand domain-containing protein D1; EFHD1; Mitocalcin; MST133; MSTP133; PP3051; RGD1559565; Swiprosin-2; SWS2 |
| Gene Symbols | EFHD1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
