missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EID2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15718831 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15718831 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15718831 Supplier Invitrogen™ Supplier No. PA561012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: AREAAFDAEY QRNPHRVDLD ILTFTIALTA SEVINPLIEE LGCDKFINRE Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

EID2 interacts with EP300 and acts as a repressor of MYOD-dependent transcription and muscle differentiation. Inhibits EP300 histone acetyltransferase activity. Acts as a repressor of TGFB/SMAD transcriptional responses. It may act as a repressor of the TGFB/SMAD3-dependent signaling by selectively blocking formation of TGFB-induced SMAD3-SMAD4 complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EID2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EID2
Gene Accession No. Q8N6I1
Gene Alias CREBBP/EP300 inhibitor 2; CREBBP/EP300 inhibitory protein 2; Cri2; E1A like inhibitor of differentiation 2; EID-1-like inhibitor of differentiation 2; Eid2; EID-2; EP300 interacting inhibitor of differentiation 2; EP300-interacting inhibitor of differentiation 2
Gene Symbols EID2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EID2. Recombinant protein control fragment (Product #RP-99182).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163126
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.