missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eIF2b beta Polyclonal Antibody

Product Code. 16529391
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16529391 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16529391 Supplier Invitrogen™ Supplier No. PA596551

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MPGSAAKGSE LSERIESFVE TLKRGGGPRS SEEMARETLG LLRQIITDHR WSNAGELMEL IRREGRRMTA AQPSETTVGN MVRRVLKIIR EEYGRLHGRS DESDQQESLH KLLTSGGLNE Positive Samples: Mouse thymus, Rat kidney.

eIF2b beta is the beta subunit of eukaryotic initiation factor-2B (EIF2B). EIF2B is involved in protein synthesis and exchanges GDP and GTP for its activation and deactivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF2b beta
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Eif2b2
Gene Accession No. P49770, Q62818, Q99LD9
Gene Alias AA409345; C85417; EIF2B; eIF-2B GDP-GTP exchange factor subunit beta; Eif2b2; EIF2BB; EIF-2Bbeta; eukaryotic translation initiation factor 2B subunit beta; eukaryotic translation initiation factor 2B, subunit 2 (beta, 39kD); eukaryotic translation initiation factor 2B, subunit 2 beta; eukaryotic translation initiation factor 2B, subunit 2 beta, 39kDa; S20I15; S20III15; translation initiation factor eIF-2B subunit beta
Gene Symbols Eif2b2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human EIF2B2 (NP_0550541).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217715, 84005, 8892
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.