Learn More
ELA3A Antibody (CELA3A/12552), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44582-100ug
This item is not returnable.
View return policy
Description
ELA3A Monoclonal antibody specifically detects ELA3A in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| ELA3A | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| CELA3A/12552 | |
| Unconjugated | |
| chymotrypsin-like elastase family member 3A, chymotrypsin-like elastase family, member 3A, EC 3.4.21, EC 3.4.21.70, ELA3A, ELA3elastase 1, elastase 3A, pancreatic, elastase 3A, pancreatic (protease E), Elastase IIIA, elastase-3A, Protease E | |
| Recombinant fragment (around aa50-250) of human ELA3A protein (Uniprot: P09093). Exact sequence is proprietary. | |
| 100 μg | |
| Lipid and Metabolism | |
| 10136 | |
| Store at 4⪚C. | |
| IgG1 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido