missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELA3A Antibody (CELA3A/12552), Novus Biologicals™
Mouse Monoclonal Antibody
256.00€ - 484.00€
Specifications
| Antigen | ELA3A |
|---|---|
| Clone | CELA3A/12552 |
| Concentration | 0.2 mg/ml |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30303182
|
Novus Biologicals
NBP3-44582-100ug |
100 μg |
484.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30295926
|
Novus Biologicals
NBP3-44582-20ug |
20 μg |
256.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ELA3A Monoclonal antibody specifically detects ELA3A in Human samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| ELA3A | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| chymotrypsin-like elastase family member 3A, chymotrypsin-like elastase family, member 3A, EC 3.4.21, EC 3.4.21.70, ELA3A, ELA3elastase 1, elastase 3A, pancreatic, elastase 3A, pancreatic (protease E), Elastase IIIA, elastase-3A, Protease E | |
| Recombinant fragment (around aa50-250) of human ELA3A protein (Uniprot: P09093). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
| CELA3A/12552 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| 10 mM PBS with 0.05% BSA | |
| 10136 | |
| IgG1 κ | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title