missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EMR2 Monoclonal Antibody (2A1)
GREENER_CHOICE

Product Code. 16312494
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16312494 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16312494 Supplier Invitrogen™ Supplier No. MA528205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse anti Human CD312 antibody, clone 2A1 recognizes human EMR2, a member of the epidermal growth factor-seven transmembrane (EGF-TM7) family of proteins, which is closely related to CD97.

EMR3 gene encodes a member of the class B seven-span transmembrane (TM7) receptor family expressed predominantly by cells of the immune system. Family members are characterized by an extended extracellular region with a variable number of N-terminal epidermal growth factor (EGF)-like domains coupled to a TM7 domain via a mucin-like spacer domain. This gene is closely linked to the gene encoding egf-like molecule containing mucin-like hormone receptor 2 on chromosome 19. This protein may play a role in myeloid-myeloid interactions during immune and inflammatory responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMR2
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene ADGRE2
Gene Accession No. Q9UHX3
Gene Alias ADGRE2; Adhesion G protein-coupled receptor E2; CD312; CD312 antigen; egf-like module containing, mucin-like, hormone receptor-like 2; EGF-like module receptor 2; EGF-like module-containing mucin-like hormone receptor-like 2; EMR2; VBU
Gene Symbols ADGRE2
Host Species Mouse
Immunogen NIH-3T3 cells stably transfected with EMR2 (EGF1-5) cDNA.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30817
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.