Learn More
Invitrogen™ Endothelin B Receptor Recombinant Rabbit Monoclonal Antibody (24GB3375)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557023
 View more versions of this product
Product Code. 30275760
  • 500.00€ / 100µL
Description
Endothelin B Receptor Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Endothelin B Receptor
Recombinant Monoclonal
0.8 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P21451, P24530, P48302
EDNRB
A synthesized peptide derived from human Endothelin B Receptor (aa 35-90).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB3375
Unconjugated
EDNRB
ABCDS; AU022549; Ednra; Ednrb; endothelin B receptor; endothelin receptor non-selective type; endothelin receptor subtype B1; endothelin receptor type B; endothelin-B receptor; ETb; ET-B; Etb receptor; Et-b receptor; ETB1; ETBR; ET-BR; Et-b-r; Etb-svr; ETRB; ET-RB; ETR-b; HSCR; HSCR2; piebald; RP11-318G21.1; Sox10m1; WS4A
Rabbit
Affinity chromatography
RUO
13618, 1910, 50672
-20°C