missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Endothelin B Receptor Recombinant Rabbit Monoclonal Antibody (24GB3375)

Rabbit Recombinant Monoclonal Antibody
146.00€ - 500.00€
Specifications
| Antigen | Endothelin B Receptor |
|---|---|
| Clone | 24GB3375 |
| Concentration | 0.8 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275760
![]() |
Invitrogen™
MA557023 |
100 μL |
500.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275308
|
Invitrogen™
MA557024 |
20 μL |
146.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Endothelin B Receptor Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.Specifications
| Endothelin B Receptor | |
| 0.8 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P21451, P24530, P48302 | |
| 13618, 1910, 50672 | |
| A synthesized peptide derived from human Endothelin B Receptor (aa 35-90). | |
| Antibody |
| 24GB3375 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| EDNRB | |
| ABCDS; AU022549; Ednra; Ednrb; endothelin B receptor; endothelin receptor non-selective type; endothelin receptor subtype B1; endothelin receptor type B; endothelin-B receptor; ETb; ET-B; Etb receptor; Et-b receptor; ETB1; ETBR; ET-BR; Et-b-r; Etb-svr; ETRB; ET-RB; ETR-b; HSCR; HSCR2; piebald; RP11-318G21.1; Sox10m1; WS4A | |
| EDNRB | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title