missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EphA10 Polyclonal Antibody, Invitrogen™

Product Code. 17978872
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17978872 200 μL 200µL
17929552 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17978872 Supplier Invitrogen™ Supplier No. PA5145427

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EPHA10 is a receptor for members of the ephrin-A family. It binds to EFNA3, EFNA4 and EFNA5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene EPHA10
Gene Accession No. Q5JZY3, Q8BYG9
Gene Alias A330090H18; EPH receptor A10; EPHA10; EphA10s protein; ephrin type-A receptor 10; RGD1562679
Gene Symbols EPHA10
Host Species Rabbit
Immunogen Recombinant Human EPHA10 protein, Val645-Leu900 (Accession #Q5JZY3).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230735, 284656, 298528
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.