missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERR beta/NR3B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB300-748-0.025mg
This item is not returnable.
View return policy
Description
ERR beta/NR3B2 Polyclonal specifically detects ERR beta/NR3B2 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ERR beta/NR3B2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 1mg/ml BSA with 0.05% Sodium Azide | |
| ESRRB | |
| Synthetic peptide corresponding to residues D(376) Y E L S Q R H E E P R R(388) of mouse ERR2. | |
| 0.025 mg | |
| Primary | |
| Estrogen-Related Receptor beta | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 2 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| estrogen-related receptor beta | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2103 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction