missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERR beta/NR3B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
189.00€
Specifications
| Antigen | ERR beta/NR3B2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
ERR beta/NR3B2 Polyclonal specifically detects ERR beta/NR3B2 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| ERR beta/NR3B2 | |
| Western Blot 2 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| estrogen-related receptor beta | |
| ESRRB | |
| IgG | |
| Affinity Purified | |
| Estrogen-Related Receptor beta |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 1mg/ml BSA with 0.05% Sodium Azide | |
| 2103 | |
| Synthetic peptide corresponding to residues D(376) Y E L S Q R H E E P R R(388) of mouse ERR2. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title