missing translation for 'onlineSavingsMsg'
Learn More

Erucic Acid Methyl-d3 Ester, TRC

Product Code. 30374819
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30374819 250 mg 250mg
30386025 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30374819 Supplier Toronto Research Chemicals Supplier No. TRCE649917250MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Erucic Acid Methyl-d3 Ester
Molecular Formula C23H41D3O2
Quantity 250 mg
Synonym (13Z)-13-Docosenoic Acid Methyl-d3 Ester, (Z)-13-Docosenoic Acid Methyl-d3 Ester, Brassidic acid methyl-d3 ester, (Z)-Erucic acid methyl-d3 ester, Agnique ME 22U-d3, Methyl-d3 13(Z)-docosenoate, Methyl-d3 cis-13-docosenoate, Methyl-d3 erucate
InChI Formula InChI=1S/C23H44O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(24)25-2/h10-11H,3-9,12-22H2,1-2H3/b11-10-/i2D3
SMILES CCCCCCCC/C=C\CCCCCCCCCCCC(OC([2H])([2H])[2H])=O
IUPAC Name methyl-d3 (Z)-docos-13-enoate
Molecular Weight (g/mol) 355.61
Formula Weight 355.35
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.