missing translation for 'onlineSavingsMsg'
Learn More

Erythritol-13C4, TRC

Product Code. 30381935
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381935 25 mg -
30373742 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381935 Supplier Toronto Research Chemicals Supplier No. TRCE65010225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C4H10O4
Molecular Weight (g/mol) 126.09
Synonym C* Eridex 16954-13C4, C*Eridex-13C4, Cargill Zerose 16957-13C4, Erythrit-13C4, Erythritol 100M-13C4, Erythrol-13C4, F 8015-13C4, Lakanto-S-13C4, Mesoerythritol-13C4, NIK 242-13C4, NSC 8099-13C4, Phycitol-13C4, Zerose-13C4, Zerose 01657-13C4, meso-Erythritol-13C4
SMILES O[13C][13C@@](O)[13C@@](O)[13C]O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C4H10O4/c5-1-3(7)4(8)2-6/h3-8H,1-2H2/i1+1,2+1,3+1,4+1
Chemical Name or Material Erythritol-13C4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.