missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,2-difluoropropionate, 97%

Product Code. 15404507
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15404507 1 g 1g
15414507 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404507 Supplier Thermo Scientific Alfa Aesar Supplier No. H31852.03
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 28781-85-3 | C5H8F2O2 | 138.114 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28781-85-3
Molecular Formula C5H8F2O2
Molecular Weight (g/mol) 138.114
MDL Number MFCD06657970
InChI Key ISVLDAIKRGXNCZ-UHFFFAOYSA-N
Synonym 2,2-difluoropropionic acid ethyl ester, ethyl 2,2-difluoropropionate, 2,2-difluoropropionicacidethylester, 2,2-difluoro-propionic acid ethyl ester, propanoic acid, 2,2-difluoro-, ethyl ester, ethyl difluoropropionate, ethyl 2,2 difluoropropionate, difluoropropionic acid ethyl ester, 2,2-difluoropropionic acidethyl ester
PubChem CID 2761188
IUPAC Name ethyl 2,2-difluoropropanoate
SMILES CCOC(=O)C(C)(F)F

Specifications

Boiling Point 24°C (20 mmHg)
Linear Formula CH3CF2CO2CH2CH3
Quantity 1 g
UN Number UN3272
Formula Weight 138.11
Percent Purity 97%
Chemical Name or Material Ethyl 2,2-difluoropropionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.